Learn More
Description
The RING-finger ubiquitin E3 ligase MDM2 (Murine Double Minute 2) is an important regulator of the tumor suppressor protein p53. MDM2 ligase activity is at least partially responsible for the ubiquitination and subsequent proteasomal degradation of p53 and other proteins.
Ubiquitinated proteins can be used in downstream applications, or analyzed by Western blot using antibodies specific for the target protein
Specifications
Specifications
| Content And Storage | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Type | ACTFS, Angiocidin, ASF, HDM2, HDMX, Macropain, Mcb1, MDM2 oncogene, E3 ubiquitin protein ligase, PSMD4, pUB-R5, Rpn10 S5a |
| Quantity | 1 Kit |
| Gene ID (Entrez) | 4193 |
| Gene Symbol | MDM2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
