Learn More
R&D Systems™ Human Myeloid Dendritic Cell Multi-Color Flow Kit
Click to view available options
:
25 Tests
Description
Single-step staining
Conjugated antibodies (and corresponding isotype controls): BDCA1/CD1c-APC (goat IgG), BDCA3/CD141-PE (Clone 501733; mouse IgG1), CD16-PerCP (Clone 245536; mouse IgG2A), CD11c-CFS (Clone ICRF 3.9; mouse IgG1)
Specifications
Specifications
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |