Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human NDST1 Protein

Catalog No. 5967ST010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to transfer sulfate from PAPS to heparan sulfate. The specific activity is >750 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P52848
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, Brij-35 and Glycerol.
Gene ID (Entrez) 3340
Molecular Weight (g/mol) M.W. (Observed): 90-100 kDa, reducing conditions; M.W. (theoretical): 93 kDa
Name N-Deacetylase/N-Sulfotransferase 1/NDST1
Quantity 10 μg
Source Chinese Hamster Ovary cell line, CHO-derived human N-Deacetylase/N-Sulfotransferase 1/NDST1 protein Ser79-Arg882, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol NDST1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein N-Deacetylase/N-Sulfotransferase 1/NDST1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.