Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Neurolysin Protein

Catalog No. 3814ZN010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave a fluorogenic peptide substrate, (7-methoxycoumarin-4-yl)acetyl-Pro-Leu-Gly-Pro-D-Lys(2,4-dinitrophenyl)-OH or Mca-PLGPK(Dnp)-OH. The specific activity is >70 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q9BYT8
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 57486
Molecular Weight (g/mol) M.W. (Observed): 73 kDa, reducing conditions; M.W. (theoretical): 77 kDa
Name Neurolysin
Quantity 10 μg
Source E. coli-derived human Neurolysin protein Ser38-Pro704, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol NLN
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Neurolysin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.