Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human PDGF R alpha Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS.
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human PDGF R alpha is immobilized at 2 μg/mL (100 μL/well), Recombinant Human PDGF-AB (Catalog # 222-AB ) binds with an ED50 of 2-12 ng/mL.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 5156 |
Molecular Weight (g/mol) | 56 kDa |
Name | PDGFR alpha |
Quantity | 50 μg |
Source | Mouse myeloma cell line,NS0-derived human PDGF R alpha protein Gln24-Glu524 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction