Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human PlGF-3 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
Description
The Recombinant Human PlGF-3 Protein is derived from Sf 21 (baculovirus). The Recombinant Human PlGF-3 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in HCl. |
Gene ID (Entrez) | 5228 |
Molecular Weight (g/mol) | 22.8 kDa (monomer) |
Name | PlGF-3 |
Quantity | 25 μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human PlGF-3 protein Leu19-Arg221 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction