Learn More
R&D Systems™ Human Pluripotent Stem Cell 3-Color Immunocytochemistry Kit
Catalog No. SC021
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
10X NL557-conjugated Goat Anti-Human SOX2, 10X NL637-conjugated Goat Anti-Human Oct-3/4, 10X NL493-conjugated Goat Anti-Human Nanog
To verify pluripotency in human stem cells using single-step ICC
Defines the starting stem cell population
Increases confidence in pluripotent status by combining 3 different fluorochrome-conjugated antibodies for simultaneous staining
Reduces variability within an experiment and between studies
Allows researchers t
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.