Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human GALNT3 Protein
Click to view available options
Quantity:
20 μg
Description
Measured by its ability to transfer GalNAc from UDP-GalNAc to peptide EA2 from AnaSpec, Inc. The specific activity is >350 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
Accession Number | Q14435 |
For Use With (Application) | Enzyme Activity |
Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
Gene ID (Entrez) | 2591 |
Molecular Weight (g/mol) | M.W. (Observed): 60-75 kDa, reducing conditions; M.W. (theoretical): 69 kDa |
Name | Polypeptide GalNac Transferase 3/GALNT3 |
Quantity | 20 μg |
Source | Mouse myeloma cell line, NS0-derived human Polypeptide GalNac Transferase 3/GALNT3 protein Gln38-Asp633, with N-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction