Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Pro-Collagen I alpha 1/COL1A1 Protein
Click to view available options
Quantity:
20 μg
Description
Measured by the cleavage of its C-terminal propeptide by Recombinant Human BMP-1/PCP (Catalog # 1927-ZN ). >50% of full-length Pro-Collagen I alpha 1 is cleaved by Recombinant Human BMP-1/PCP (Catalog # 1927-ZN ), as measured under the described conditions.
Specifications
Specifications
Accession Number | P02452 |
For Use With (Application) | Enzyme Activity |
Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
Gene ID (Entrez) | 1277 |
Molecular Weight (g/mol) | M.W. (Observed): 79-83 kDa, 54-56 kDa, 35-36 kDa, reducing conditions; M.W. (theoretical): 65 kDa (full-length), 40 kDa, 28 kDa |
Name | Pro Collagen I alpha 1 |
Quantity | 20 μg |
Source | Chinese Hamster Ovary cell line, CHO-derived human Collagen I alpha 1 protein Gln23-Lys277 + Gly1094-Leu1464, with an N-terminal 6-His tag. |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction