Learn More
Description
Includes
Human proinsulin microplate, 96-well; Human proinsulin conjugate; Human proinsluin standard; Buffers and diluents, Color reagents A and B; Stop solution; Plate sealers
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | HRP |
| Assay Sensitivity | 1.43 pM |
| Assay Range | 3.28 to 210 pM |
| For Use With (Application) | ELISA |
| Product Line | R&D Systems |
| Sample Type | Serum (100uL), EDTA Plasma (100uL), Heparin Plasma (100uL) |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
