Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Proprotein Convertase 9/PCSK9 Protein

Catalog No. 3888se010
Click to view available options
Quantity:
10 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Human LDL R (Catalog # 2148-LD ) is coated at 2 μg/mL, Recombinant Human Proprotein Convertase 9/PCSK9 binds with an ED50 of 70-420 ng/mL.

Specifications

Accession Number Q8NBP7
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 255738
Molecular Weight (g/mol) M.W. (Observed): 19 kDa & 66 kDa, reducing conditions; M.W. (theoretical): 14 kDa & 59 kDa
Name Proprotein Convertase 9/PCSK9
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Proprotein Convertase 9/PCSK9 protein Gln31-Gln152 & Ser153-Gln692, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Mature form & pro domain
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9
Gene Symbol PCSK9
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Proprotein Convertase 9/PCSK9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.