Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Protein Fraction II Protein Fraction

Catalog No. p-4720655
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Source: Human cervical epithelial carcinoma cell line, HeLa-derived
Formulation: Supplied as a solution in HEPES.
Regulatory Status: RUO

Specifications

Formulation Supplied as a solution in HEPES.
Name Protein Fraction II
Quantity 1 mg
Conjugate Unconjugated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.