Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human SCGF-alpha/CLEC11a Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Description
The Recombinant Human SCGF-alpha/CLEC11a Protein is derived from NS0. The Recombinant Human SCGF-alpha/CLEC11a Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Accession Number | Q9Y240 |
Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Gene ID (Entrez) | 6320 |
Molecular Weight (g/mol) | 34.7 kDa |
Name | SCGF/CLEC11a |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived human SCGF/CLEC11a protein Gly19-Phe323,with an N-terminal Met and a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Gene Alias | CLEC11a, CLECSF3, LSLCL, P47, SCGF |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction