Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Sphingosine Kinase 1/SPHK1 Protein

Catalog No. 5536SK010
Click to view available options
:
10ug; Carrier Free
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Sphingosine Kinase 1/SPHK1 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Sphingosine Kinase 1/SPHK1 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation Supplied as a 0.2 μm filtered solution in MES, NaCl, Glycerol, DTT and Triton™ .
Gene ID (Entrez) 8877
Molecular Weight (g/mol) 45 kDa
Name Sphingosine Kinase 1/SPHK1
Quantity 10μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Sphingosine Kinase 1/SPHK1 protein Asp2-Leu398 with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >75%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.