Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human SUMO1 Biotin Recombinant Protein

Catalog No. ul725050
Click to view available options
:
50ug; Carrier Free
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human SUMO1 Biotin Protein is derived from E. coli. The Recombinant Human SUMO1 Biotin Protein has been validated for the following applications: Enzyme Activity.

Specifications

Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 7341
Name SUMO1 Biotin
Quantity 50μg
Source E. coli-derived human SUMO1 protein
Storage Requirements Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Conjugate Biotin
Recombinant Recombinant
Purity or Quality Grade 95%, by PAGE.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.