Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Ubiquitin Fluorescein Recombinant Protein
Click to view available options
Quantity:
100 μg
Description
The Recombinant Human Ubiquitin Fluorescein Protein is derived from E. coli. The Recombinant Human Ubiquitin Fluorescein Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | No Formulation Info in Bulk Inventory |
Gene ID (Entrez) | 7314 |
Name | Ubiquitin Fluorescein |
Quantity | 100 μg |
Source | E. coli-derived human Ubiquitin protein |
Conjugate | Fluorescein |
Recombinant | Recombinant |
Purity or Quality Grade | 95%, by PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction