Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF 165 Protein
GSA_VA

Catalog No. 293VE001MGC
Click to view available options
Quantity:
1 mg
10 μg
50 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1-6 ng/mL. The specific activity of recombinant human VEGF165 is approximately 1.7 x 103 U/μg, which is calibrated against recombinant human VEGF165 WHO Standard (NIBSC code: 02/286). Specific activity is for reference purposes only and is not routinely tested.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) observed_molecular_weight: 20-22 kDa, under reducing conditions. 39-42 kDa, under non-reducing conditions.
Name VEGF 165
Quantity 1 mg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS. Alternatively, reconstitute (at 100-500 μg/mL in sterile 4 mM HCl.)
Purity or Quality Grade 97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.