Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Influenza A Virus H3N2 Neuraminidase Protein
Click to view available options
Quantity:
5 μg
Description
Measured by its ability to cleave a fluorogenic substrate, 2’-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid. The specific activity is >10,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
Accession Number | Q91MA2 |
For Use With (Application) | Enzyme Activity |
Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2, DTT and Glycerol. |
Molecular Weight (g/mol) | M.W. (Observed): 57-65 kDa, reducing conditions; M.W. (theoretical): 53 kDa |
Name | Viral Neuraminidase |
Quantity | 5 μg |
Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived influenza a virus h3n2 Viral Neuraminidase protein Ala46-Ile469, vasodilator-stimulated phosphoprotein tetramerization domain and a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction