Learn More
R&D Systems™ MagCellect CD24- CD44+ Breast Cancer Stem Cell Isolation Kit
SDP
Catalog No. MAGH111 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
MAGH111 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. MAGH111 Supplier R&D Systems™ Supplier No. MAGH111
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolation by positive and negative selection improves purity of recovered cells
Cancer stem cells are a rare population of self-renewing cells found in tumors and in cancer cell lines. Methods that use positive or negative selection to enrich for cancer stem cells can lead to poor recovery and low cell purity as well as the loss of valuable tissue samples and resources. Methods that use both positive and negative selection to isolate cancer stem cells can yield cells with a higher purity.
Isolation based on two markers increases the purity of recovered cells
Isolates CD24low/- CD44+ cells which have been shown to form tumors in mice
Includes fluorochrome-conjugated CD24 and CD44 antibodies for analysis of recovered cells
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit