Learn More
Description
CD14 is a 55kDa GPI-linked cell surface glycoprotein that is preferentially expressed on monocytes/macrophages. The amino acid sequence of mature human CD14 is approximately 67% and 64% identical to the mouse and rat proteins, respectively.
Typical recovery ranges between 45% and 75% and the purity of recovered CD14+ cells ranges between 90 and 97%
Monocytes isolated using the MagCellect CD14+ Cell Isolation Kit can be used for various downstream applications including the ex vivo generation of dendritic cells
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Kit |
| Type | Cell Isolation Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
