Description
The resulting cell preparation is highly enriched for CD4+CD25+ T cells with a purity of recovered cells ranging between 85-95%
Two-step procedure
Includes Streptavidin Ferrofluid for both the negative selection step to isolate CD4+ T cells, and in the positive selection step to isolate CD4+CD25+ regulatory T cells
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Kit |