Learn More
R&D Systems™ MagCellect Mouse Memory CD4+ T Cell Isolation Kit
SDP
Catalog No. MAGM206 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
MAGM206 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. MAGM206 Supplier R&D Systems™ Supplier No. MAGM206
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Designed to isolate Memory CD4+ T cells by negative selection
CD4 is an approximately 55kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T cells. In humans, CD4 is also expressed to a lesser extent on monocytes and macrophage related cells.
Typical recoveries range between 5 and 35% and the purity of recovered cells ranges between 85 and 95%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit