Description
Typical recovery of the targeted cell population ranges between 40% and 65% and the purity of recovered B cells ranges from 89-95%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
Magnetically tagged cells will migrate toward the magnet (unwanted cell fraction)
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 1 Kit |