Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mca-PLGL-Dpa-AR-NH2 Fluorogenic MMP Substrate

Catalog No. es001
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Applications: Bioactivity, Binding Activity

  • Substrate for MMP-1, MMP-2, MMP-7, MMP-8, MMP-9, MMP-12, MMP-13, MMP-14, MMP-15, MMP-16, Cathepsin D, and Cathepsin E
  • Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
  • It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
  • Also an excellent substrate for active Cathepsins D and E
  • Purity 98±1% based on HPLC

Specifications

Form Liquid
For Use With (Application) Bioactivity, Binding Activity
Quantity 1 mg
Format Supplied as a stock solution in dimethyl sulfoxide (DMSO) at a concentration of 7.99mg/mL or 6.07 mM.
Molecular Weight (g/mol) Predicted MW: 1093 Da
Product Type Source: Synthetic
Storage Requirements Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.