Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ML 179
GREENER_CHOICESDP

Catalog No. 495750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
495750 50 mg
495710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 495750 Supplier Tocris Bioscience™ Supplier No. 4957/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective liver receptor homolog 1 (LRH1) inverse agonist

ML 179 is a selective liver receptor homolog 1 (LRH1, NR5A2) inverse agonist (IC50 = 320 nM and maximum efficacy of 64% repression). Exhibits >31-fold selectivity over SF1.

Specifications

Chemical Name or Material ML 179
CAS 1883548-87-5
Synonym 3-Cyclohexyl-6-[4-[3-(trifluoromethyl)phenyl]piperazin-1-yl]-1H-pyrimidine-2,4-dione
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C21H25F3N4O2
Formula Weight 422.44
Physical Form Crystalline Solid
Product Type Bioactive Small Molecule
Purity >99%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.