Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Carboxylesterase 2/CES2 Protein
Click to view available options
:
10ug; Carrier Free
Description
Measured by its ability to hydrolyze p-nitrophenylacetate. The specific activity is >35,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q91WG0 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 8824 |
| Molecular Weight (g/mol) | M.W. (Observed): 58 kDa, reducing conditions; M.W. (theoretical): 60 kDa |
| Name | Carboxylesterase 2/CES2 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse Carboxylesterase 2/CES2 protein Gln27-Lys557, with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction