Description
CD3+ is a universal marker for T cells.
Based upon gravitational flow
Simple procedure
No specialized equipment required
Specifications
Specifications
| Description | Mouse CD3+ T Cell Enrichment Column, Large |
| For Use With (Application) | Prepare purified murine T cell populations via high affinity negative selection |
| Product Line | R&D Systems |
| Quantity | 1 Column |