Learn More
Description
CD3+ is a universal marker for T cells.
Total CD3+ cell recovery from all available CD3+ cells loaded range between 43% and 51%
Purity of recovered cells ranges between 83% and 90%
Based upon gravitational flow
Simple procedure
No specialized equipment required
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Includes | Murine T Cell Selection Columns, 10X Column Buffer Concentrate |
| Product Line | R&D Systems |
| Quantity | 25/Pk. |
| Type | Cell Enrichment Column |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
