Learn More
R&D Systems™ Mouse CD4+/CD62L-/CD44 high Memory Columns
SDP
Catalog No. MCD44 Shop All Novus Products
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
MCD44 4/Pk.
1 options
This item is not returnable. View return policy
Catalog No. MCD44 Supplier R&D Systems™ Supplier No. MCD44
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Designed to prepare sub-populations of CD4 mouse memory T cells via high affinity negative selection
Recovery of CD4+/CD62L-/CD44high memory T cells from all available CD4+/CD44+ T cells loaded ranges from 5% to 23% and is dependent upon the strain of mouse used for study
The purity of recovered cells ranges between 85 and 95%
Leukocyte suspensions are incubated with a mixture of monoclonal antibodies and then loaded onto T Cell Columns
B cells, nonselected T cells, and monocytes bind to glass beads coated with anti-immunoglobulin via both F(ab) and Fc interactions
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 4/Pk.