Learn More
R&D Systems™ Mouse CD4+/CD62L+/CD44 low Naive Columns
SDP
Catalog No. MCD45 Shop All Novus Products
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
MCD45 4/Pk.
1 options
This item is not returnable. View return policy
Catalog No. MCD45 Supplier R&D Systems™ Supplier No. MCD45
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Designed to prepare sub-populations of CD4 mouse naive T cells via high affinity negative selection
Recovery of CD4+/CD62L+/CD44low naive T cells from all available CD4+/CD62L+ cells loaded ranges between 5% and 23% and is dependent upon the strain of mouse used for study
The purity of the recovered cells
ranges between 85 and 95%
Leukocyte suspensions are incubated with a mixture of monoclonal antibodies and then loaded onto T Cell Columns
B cells, nonselected T cells, and monocytes bind to glass beads coated with anti-immunoglobulin via both F(ab) and Fc interactions
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 4/Pk.