Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Dectin-2/CLEC6A Protein
Click to view available options
Quantity:
50 μg
Description
The Recombinant Mouse Dectin-2/CLEC6A Protein is derived from NS0. The Recombinant Mouse Dectin-2/CLEC6A Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 56620 |
Molecular Weight (g/mol) | 20.6 kDa |
Name | Dectin-2/CLEC6A α Isoform |
Quantity | 50 μg |
Source | Mouse myeloma cell line,NS0-derived mouse Dectin-2/CLEC6A protein Phe43-Leu209,with an N-terminal 7-His tag and Ala |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction