Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse EG-VEGF/PK1 Protein

Catalog No. p-4721413
Click to view available options
Formulation:
Supplied as a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse EG-VEGF/PK1 Protein is derived from NS0. The Recombinant Mouse EG-VEGF/PK1 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 246691
Molecular Weight (g/mol) 9.6 kDa
Name EG-VEGF/PK1
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse EG-VEGF/PK1 protein Ala20-Phe105
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.