Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse G-CSF Protein
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20 with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20 with Trehalose.
Description
The Recombinant Mouse G-CSF Protein is derived from E. coli. The Recombinant Mouse G-CSF Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in Citric Acid and Tween™ 20 with Trehalose. |
Gene ID (Entrez) | 12985 |
Molecular Weight (g/mol) | 19 kDa |
Name | G-CSF |
Quantity | 5 μg |
Source | E. coli-derived mouse G-CSF protein Val31-Ala208,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction