Learn More
R&D Systems™ Mouse Hematopoietic Stem Cell Expansion Cytokine Panel
Catalog No. smpk9
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
100μg each of recombinant mouse Flt-3 Ligand, Tpo, and SCF
Sufficient for 1 liter of media
The combined use of multiple cytokines, Flt-3 ligand, SCF and Tpo, permits the ex vivo expansion of hematopoietic stem cells (HSC)
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.