Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse IBSP/Sialoprotein II Protein
Click to view available options
Quantity:
50 μg
Description
The Recombinant Mouse IBSP/Sialoprotein II Protein is derived from CHO. The Recombinant Mouse IBSP/Sialoprotein II Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 15891 |
Molecular Weight (g/mol) | 34.9 kDa |
Name | IBSP/Sialoprotein II |
Quantity | 50 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse IBSP/Sialoprotein II protein Met1-Gln324,with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction