Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse IFN-gamma Protein

Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol,Tween™ 80 and TCEP with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol,Tween™ 80 and TCEP with Trehalose.
Description
The Recombinant Mouse IFN-gamma Protein is derived from E. coli. The Recombinant Mouse IFN-gamma Protein has been validated for the following applications: Bioactivity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol,Tween™ 80 and TCEP with BSA as a carrier protein. |
Gene ID (Entrez) | 15978 |
Molecular Weight (g/mol) | 15.6 kDa |
Name | IFN-gamma |
Quantity | 100 μg |
Source | E. coli-derived mouse IFN-gamma protein His23-Cys155,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction