Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-10 R alpha Recombinant Protein

Catalog No. 474MR025
Click to view available options
Quantity:
25 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

A New and Improved rm IL-10 Ra is Now Available! It has a lower endotoxin specification!

Measured by its ability to inhibit IL-10-dependent proliferation of MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. Approximately 1-3 µg/mL of recombinant mouse IL-10 sR alpha will inhibit 50% of the biological response due to 1 ng/mL of recombinant mouse IL-10.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 16154
Molecular Weight (g/mol) 24 kDa
Name IL-10 R alpha
Quantity 25 μg
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.