Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-15 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 20.
Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein.
Description
The Recombinant Mouse IL-15 Protein is derived from E. coli. The Recombinant Mouse IL-15 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 20. |
Gene ID (Entrez) | 16168 |
Molecular Weight (g/mol) | 13.4 kDa |
Name | IL-15 |
Quantity | 10 μg |
Source | E. coli-derived mouse IL-15 protein Asn49-Ser162,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction