Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-2 Protein

Catalog No. 402ML100CF
Click to view available options
Quantity:
100 μg
20 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80 with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-2 Protein is derived from E. coli. The Recombinant Mouse IL-2 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80.
Gene ID (Entrez) 16183
Molecular Weight (g/mol) 17.2 kDa
Name IL-2
Quantity 100 μg
Source E. coli-derived mouse IL-2 protein Ala21-Gln169,with and without an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100-200μg/mL in sterile deionized water.
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.