Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-36 gamma/IL-1F9 (aa 13-164) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in MES, NaCl, TCEP, EDTA, CHAPS and PEG 8000 with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP, EDTA and Trehalose.
Description
The Recombinant Mouse IL-36 gamma/IL-1F9 (aa 13-164) Protein is derived from E. coli. The Recombinant Mouse IL-36 gamma/IL-1F9 (aa 13-164) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MES, NaCl, TCEP, EDTA, CHAPS and PEG 8000 with BSA as a carrier protein. |
| Gene ID (Entrez) | 215257 |
| Molecular Weight (g/mol) | 17.3 kDa |
| Name | IL-36 gamma/IL-1F9 (aa 13-164) |
| Quantity | 10 μg |
| Source | E. coli-derived mouse IL-36 gamma/IL-1F9 protein Gly13-Ser164 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Monomer |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction