Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha 10 beta 1 Protein
Click to view available options
:
50ug; Carrier Free
Description
The Recombinant Mouse Integrin alpha 10 beta 1 Protein is derived from CHO. The Recombinant Mouse Integrin alpha 10 beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Molecular Weight (g/mol) | 129.2 kDa (Integrin alpha10) & 86.5 kDa (Integrin beta1) |
Name | Integrin alpha 10 beta 1 |
Quantity | 50μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 10 beta 1 protein, (N-terminus) Mouse Integrin alpha 10 Phe23-Thr1118 (Lys ins between Gln858 and Ala859) Accession &Num; NP_001289400.1 His-Pro GGGSGGGS Acidic Tail HHHHHH Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055 His-Pro GGGSGGGS Basic Tail (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Noncovalently-linked heterodimer |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction