Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Kininogen HMW (HKa) Protein
Click to view available options
Quantity:
10 μg
Description
The Recombinant Mouse Kininogen HMW (HKa) Protein is derived from NS0. The Recombinant Mouse Kininogen HMW (HKa) Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 16644 |
| Molecular Weight (g/mol) | 72 kDa (mature), 38 kDa (heavy) & 31 kDa (light) |
| Name | Kininogen High Molecular Weight (HKa) |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Kininogen protein Glu21-Ser661,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction