Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Lefty-1 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Description
The Recombinant Mouse Lefty-1 Protein is derived from NS0. The Recombinant Mouse Lefty-1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 10637 |
Molecular Weight (g/mol) | 26.9 kDa |
Name | Lefty-1 |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived mouse Lefty-1 protein Mouse Lefty-1 Proregion (Leu22-Arg135) Accession &Num; NP_034224 6-His tag Mouse Lefty mature (Leu136-Pro368) Accession &Num; NP_034224 6-His tag Mouse Lefty mature (Leu136-Pro368) Accession &Num; NP_034224 N-terminus C-terminus A short DNA sequence encoding 6 histidine residues was inserted into the DNA sequence encoding mouse Lefty-1 between Arg135 and Leu136. Two peptides corresponding to mouse Lefty-1 propeptide and mature mouse Lefty-1 are present in the recombinant preparation. |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction