Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Nephrin Protein
Click to view available options
Quantity:
50 μg
Description
Measured by the ability of the immobilized protein to support the adhesion of MS-1 mouse pancreatic islet endothelial cells. When 5 x 104 cells/well are added to rmNephrin coated plates (10 μg/mL, 100 μL/well), >60% will adhere after 90 minutes at 37° C.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 54631 |
| Molecular Weight (g/mol) | 110 kDa |
| Name | Nephrin |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Nephrin protein Gln37-Thr1049,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction