Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Noggin Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Description
The Recombinant Mouse Noggin Protein is derived from NS0. The Recombinant Mouse Noggin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Gene ID (Entrez) | 18121 |
Molecular Weight (g/mol) | 26 kDa (monomer) |
Name | Noggin |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived mouse Noggin protein, (N-terminus) Mouse Noggin (Leu20-Cys232) Accession &Num; P97466 IEGRGGGSGGGSGGGS 10-His tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Gene Alias | NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction