Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Pentraxin 3/TSG-14 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 .
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein.
Description
The Recombinant Mouse Pentraxin 3/TSG-14 Protein is derived from NS0. The Recombinant Mouse Pentraxin 3/TSG-14 Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 . |
Gene ID (Entrez) | 19288 |
Molecular Weight (g/mol) | 41 kDa (monomer) |
Name | Pentraxin 3/TSG-14 |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived mouse Pentraxin 3/TSG-14 protein Glu18-Ser381,with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Multimer consisting of as many as ten non-covalently and covalently linked subunits |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction