Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse SP-D Quantikine ELISA Kit
SDP

Catalog No. MSFPD0 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSFPD0 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MSFPD0 Supplier R&D Systems™ Supplier No. MSFPD0

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse SP-D Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse SP-D Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse SP-D in cell culture supernates (50 ul), tissue lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Product Type ELISA Kit
Quantity 1 Kit
Species Mouse
Conjugate HRP
Assay Sensitivity 9.43 pg/mL
Assay Range 62.50 to 4000 pg/mL
For Use With (Application) ELISA
Product Line R&D Systems
Sample Type Cell Culture Supernates (50uL), Tissue Lysates (50uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL)
Synonym COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4, SFTP4pulmonary surfactant-associated protein D, SFTPD, SPD, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Immunology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.