Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse TNF-alpha ELISpot Kit
Click to view available options
Quantity:
1 x 96-well Plate
6 x 96-well Plates
Specifications
Specifications
Includes | One 96-well PVDF-backed plate coated w/pAb for mouse TNF-a, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT, Positive control |
Quantity | 1 x 96-well Plate |
Sample Type | Whole Cells |
Synonym | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Use With (Application) | Diagnostics |
Product Type | Mouse TNF-alpha ELISpot Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction