Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse VCAM-1/CD106 Fc Chimera Protein
Click to view available options
Quantity:
200 μg
50 μg
Description
The Recombinant Mouse VCAM-1/CD106 Fc Chimera Protein is derived from NS0. The Recombinant Mouse VCAM-1/CD106 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 22329 |
Molecular Weight (g/mol) | 101.9 kDa (monomer) |
Name | VCAM-1/CD106 Fc Chimera |
Quantity | 200 μg |
Source | Mouse myeloma cell line,NS0-derived mouse VCAM-1/CD106 protein, (N-terminus) Met Mouse VCAM-1 (Phe25-Glu698) Accession &Num; CAA47989 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction