Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse ZNRF3 Fc Chimera Protein

Catalog No. 8328rf050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse ZNRF3 Fc Chimera Protein is derived from NS0. The Recombinant Mouse ZNRF3 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q5SSZ7
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 407821
Molecular Weight (g/mol) 45kDa
Name ZNRF3 Fc Chimera
Preparation Method Reconstitute at 100μg/mL in PBS.
Quality Control Testing SDS-PAGE: 51-60kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse ZNRF3 protein, (N-terminus) Mouse ZNRF3 (Lys53-Met216) Accession &Num; Q5SSZ7 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Lys53
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.